Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BANK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8005
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8005 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8005 Supplier R&D Systems Supplier No. AF8005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

BANK1 Polyclonal specifically detects BANK1 in Human samples. It is validated for Western Blot.

Specifications

Antigen BANK1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q8NDB2
Gene Alias BANKFLJ20706B-cell scaffold protein with ankyrin repeats, B-cell scaffold protein with ankyrin repeats 1, FLJ34204
Gene Symbols BANK1
Host Species Sheep
Immunogen E. coli-derived recombinant human BANK1 Ser480-His785 (Cys650Arg) Accession # Q8NDB2
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55024
Test Specificity Detects human BANK1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.