Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BASP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6479SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6479SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6479SP Supplier R&D Systems Supplier No. AF6479SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

BASP1 Polyclonal specifically detects BASP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen BASP1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P80723
Gene Alias brain abundant, membrane attached signal protein 1,22 kDa neuronal tissue-enriched acidic protein, brain acid-soluble protein 1, CAP23, CAP-23, CAP23brain acid soluble protein 1, NAP22, NAP-22, NAP22MGC8555, Neuronal axonal membrane protein NAP-22, neuronal tissue-enriched acidic protein
Gene Symbols BASP1
Host Species Sheep
Immunogen E. coli-derived recombinant human BASP1 Gly2-Ala45 Accession # P80723
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10409
Test Specificity Detects human BASP1 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.