Learn More
Human BATF Alexa Fluor 750-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | BATF |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 687706 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 750 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | Q16520 |
| Gene Alias | basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA2, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.