Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BATF APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC8054A
This item is not returnable.
View return policy
Description
BATF Monoclonal specifically detects BATF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
BATF | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA2, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10538 | |
Human | |
Purified |
Flow Cytometry | |
687706 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Q16520 | |
BATF | |
E. coli-derived recombinant human BATF Met1-Ser125 Accession # Q16520 | |
100 Tests | |
Primary | |
Detects Human BATF in ELISA and Western Blot. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction