Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BATF3 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC7437G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7437G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7437G Supplier R&D Systems Supplier No. IC7437G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BATF3 Monoclonal specifically detects BATF3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen BATF3
Applications Flow Cytometry
Classification Monoclonal
Clone 841702
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Gene Accession No. Q9NR55
Gene Alias basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT, SNFT21-kD small nuclear factor isolated from T cells
Gene Symbols BATF3
Host Species Mouse
Immunogen E. coli-derived recombinant human BATF3 Met1-Arg127 Accession # Q9NR55
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 55509
Test Specificity Detects human BATF3 in ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.