Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BATF3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7437SP
This item is not returnable.
View return policy
Description
BATF3 Monoclonal specifically detects BATF3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
BATF3 | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q9NR55 | |
BATF3 | |
E. coli-derived recombinant human BATF3 Met1-Arg127 Accession # Q9NR55 | |
25 μg | |
Primary | |
Detects human BATF3 in ELISA. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
841702 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT, SNFT21-kD small nuclear factor isolated from T cells | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
55509 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction