Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Bax Minus C-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB820
Change view
Click to view available options
Quantity:
400 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB820 400 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB820 Supplier R&D Systems Supplier No. AB820
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Bax Polyclonal specifically detects Bax in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Bax
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 4 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q07812
Gene Alias apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4
Gene Symbols BAX
Host Species Goat
Immunogen Human Bax synthetic peptide
Purification Method Affinity Purified
Quantity 400 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 581
Test Specificity Detects human Bax Minus C-Terminus. Does not detect mouse Bax.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.