Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BCAM Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1481G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1481G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1481G100 Supplier R&D Systems Supplier No. FAB1481G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BCAM/CD239
Applications Flow Cytometry
Classification Monoclonal
Clone 87207
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. CAA58449
Gene Alias antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19
Gene Symbols BCAM
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BCAM Glu32-Ala547 Accession # CAA58449
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4059
Test Specificity Detects human BCAM in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhEpCAM, recombinant mouse (rm) MAdCAM-1, rhMCAM, rhNCAM-L1, rmOCAM, or rmTROP-2 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.