Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Bcl9-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF4967SP
This item is not returnable.
View return policy
Description
BCL9-2 Polyclonal specifically detects BCL9-2 in Human samples. It is validated for Western Blot.Specifications
| BCL9-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell CLL/lymphoma 9-like, B-cell CLL/lymphoma 9-like protein, B-cell lymphoma 9-like protein, Bcl92, BCL9-like protein, DLNB11, DLNB11BCL9-2, nuclear co-factor of beta-catenin signalling, Protein BCL9-2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 283149 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q86UU0 | |
| BCL9L | |
| E. coli-derived recombinant human Bcl9-2 Met38-Val206 Accession # Q86UU0 | |
| 25 μg | |
| Primary | |
| Detects endogenous human Bcl9-2 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction