Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BCMA/TNFRSF17 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1933V100UG
This item is not returnable.
View return policy
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.Specifications
BCMA/TNFRSF17 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 | |
Mouse myeloma cell line NS0-derived recombinant human BCMA/TNFRSF17, Met1-Ala54, Accession # Q02223 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Flow Cytometry | |
1039701 | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q02223 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
608 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction