Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BCMA/TNFRSF17 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB1931T100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB1931T100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB1931T100U Supplier R&D Systems Supplier No. FAB1931T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BCMA/TNFRSF17
Applications Flow Cytometry
Classification Monoclonal
Clone 1004023
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Gene Accession No. Q02223
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BCMA/TNFRSF17, Met1-Ala54, Accession No. Q02223
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 608.0
Test Specificity Detects human BCMA/TNFRSF17 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.