Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BCMA/TNFRSF17 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB193A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB193A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB193A Supplier R&D Systems Supplier No. FAB193A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

BCMA/TNFRSF17 Polyclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BCMA/TNFRSF17
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q6PE46
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17
Gene Symbols TNFRSF17
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human BCMA/TNFRSF17 Met1-Ala54 Accession # Q6PE46
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 608
Test Specificity Detects human BCMA in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhTACI, and recombinant mouse BCMA is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.