Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BDNF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF248SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF248SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF248SP Supplier R&D Systems Supplier No. AF248SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chicken Polyclonal Antibody has been used in 5 publications

BDNF Polyclonal specifically detects BDNF in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen BDNF
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 2.5 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P23560
Gene Alias abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Gene Symbols BDNF
Host Species Chicken
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF His129-Arg247 Accession # P23560
Purification Method Antigen Affinity-purified from egg yolks
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 627
Test Specificity Detects human BDNF in direct ELISAs.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.