Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-1,4-Galactosyltransferase 1/B4GalT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | beta-1,4-Galactosyltransferase 1/B4GalT1 |
---|---|
Clone | 394709 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB36091SP
![]() |
R&D Systems
MAB36091SP |
25 μg |
Each for $200.50
|
|
|||||
MAB36091100
![]() |
R&D Systems
MAB36091100 |
100 μg |
Each for $514.50
|
|
|||||
Description
beta-1,4-Galactosyltransferase 1/B4GalT1 Monoclonal specifically detects beta-1,4-Galactosyltransferase 1/B4GalT1 in Human samples. It is validated for Immunocytochemistry.Specifications
beta-1,4-Galactosyltransferase 1/B4GalT1 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P15291 | |
2683 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
394709 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
1,4- galactosyltransferase, polypeptide 1, B4GAL-T1, beta-1,4-galactosyltransferase 1, Beta-1,4-GalTase 1, Beta4Gal-T1, betaGlcNAc beta, CDG2D, EC 2.4.1, GGTB2, GGTB2DKFZp686N19253, glycoprotein-4-beta-galactosyltransferase 2, GT1, GTB, lactose synthase, MGC50983, UDP-Gal, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1 | |
Mouse myeloma cell line NS0-derived recombinant human beta-1,4-Galactosyltransferase 1/B4GalT1 Ala44-Ser398 Accession # P15291 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human beta-1,4-Galactosyltransferase 1/B4GalT1in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title