Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human beta-1,4-Galactosyltransferase 1/B4GalT1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB228768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB228768 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB228768 Supplier Novus Biologicals Supplier No. NBP280341

Specifications

Assay Range 156.25-10000 pg/mL
Assay Sensitivity 78.00 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates
Specificity Recognizes both recombinant and natural Human beta-1,4-Galactosyltransferase 1/B4GalT1.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 2683
Gene Symbol B4GALT1
Quantity 1 Kit
Research Discipline Lipid and Metabolism
Storage Requirements Storage is content dependent.
Synonym 1,4- galactosyltransferase, polypeptide 1, B4GAL-T1, beta-1,4-galactosyltransferase 1, Beta-1,4-GalTase 1, Beta4Gal-T1, betaGlcNAc beta, CDG2D, EC 2.4.1, GGTB2DKFZp686N19253, glycoprotein-4-beta-galactosyltransferase 2, GT1, GTB, lactose synthase, MGC50983, UDP-Gal, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.