Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-1,4-Glucuronyltransferase 1/B4GAT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6664SP
This item is not returnable.
View return policy
Description
Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 Monoclonal specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 in Human samples. It is validated for Western Blot.Specifications
Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
O43505 | |
B4GAT1 | |
Chinese hamster ovary cell line CHO-derived recombinant human beta-1,4-Glucuronyltransferase 1/B4GAT1 Antibody Asp43-Cys415 Accession # O43505 | |
25 μg | |
Primary | |
Detects human beta-1,3-N-acetylglucosaminyltransferase 1/B3GNT1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) B3GNT2, rhB3GNT6, or rhB4GalT1 is observed. | |
Human | |
Purified |
Western Blot | |
724057 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B3GNT1, B3GN-T1, beta-1,3-N-acetylglucosaminyltransferase bGnT-6, BETA3GNTI, EC 2.4.1.149, i-beta-1,3-N-acetylglucosaminyltransferase, iGAT, IGNT, MDDGA13, N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase, Poly-N-acetyllactosamine extension enzyme, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1B3GNT6, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11041 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction