Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human beta-1 Adrenergic R/ADRB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10119SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10119SP 25 μg
MAB10119100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10119SP Supplier R&D Systems Supplier No. MAB10119SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

beta-1 Adrenergic R/ADRB1 Monoclonal specifically detects beta-1 Adrenergic R/ADRB1 in Human samples. It is validated for Western Blot.

Specifications

Antigen beta-1 Adrenergic R/ADRB1
Applications Western Blot
Classification Monoclonal
Clone 2203D
Conjugate Unconjugated
Dilution Western Blot 1 μ/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P08588
Gene Alias ADRB1, ADRB1R, ADRB1RRHR, adrenergic, beta-1-, receptor, B1AR, beta-1 adrenergic receptor, beta1 AdrenergicR, beta-1 AdrenergicR, Beta-1 adrenoceptor, Beta-1 adrenoreceptor, BETA1AR
Host Species Rabbit
Immunogen Synthetic peptide containing human beta-1 Adrenergic R/ADRB1, Accession No. P08588
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 153
Test Specificity Detects human beta-1 Adrenergic R/ADRB1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.