Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-3 Adrenergic R/ADRB3 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9419N100UG
This item is not returnable.
View return policy
Description
beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry.Specifications
beta-3AdrenergicR/ADRB3 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
ADRB3, ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, beta3 AdrenergicR, beta-3 AdrenergicR, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960 | |
Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945 | |
100 μg | |
155 | |
Human | |
Supernatant |
Flow Cytometry | |
493324 | |
Flow Cytometry 0.25-1μg/106 cells | |
P13945 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Stains human beta-3 Adrenergic R/ADRB3 transfectants but not irrelevent transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction