Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human beta-3 Adrenergic R/ADRB3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9419SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9419SP 25 μg
MAB9419100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9419SP Supplier R&D Systems Supplier No. MAB9419SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-3 Adrenergic R/ADRB3 Monoclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen beta-3 Adrenergic R/ADRB3
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 493324
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P13945
Gene Alias ADRB3, ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, beta3 AdrenergicR, beta-3 AdrenergicR, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3 Met1-Ser408 Accession # P13945
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155
Test Specificity Stains human beta-3 Adrenergic R/ADRB3 transfectants but not irrelevent transfectants in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.