Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human beta-Arrestin 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB45601SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB45601SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB45601SP Supplier R&D Systems Supplier No. MAB45601SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

beta-Arrestin 1 Monoclonal specifically detects beta-Arrestin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen beta-Arrestin 1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 425817
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P49407
Gene Alias ARB1, ARR1, ARRB1, arrestin 2, Arrestin beta-1, arrestin, beta 1, betaArrestin 1, beta-arrestin-1
Gene Symbols ARRB1
Host Species Rat
Immunogen E. coli-derived recombinant human beta-Arrestin 1 Met1-Arg418 Accession # P49407
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 408
Test Specificity Detects human beta-Arrestin 1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.