Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-Arrestin 1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | beta-Arrestin 1 |
---|---|
Concentration | LYOPH |
Dilution | Immunohistochemistry 5-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Description
beta-Arrestin 1 Polyclonal specifically detects beta-Arrestin 1 in Human samples. It is validated for Immunohistochemistry.Specifications
beta-Arrestin 1 | |
Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
ARB1, ARR1, ARRB1, arrestin 2, Arrestin beta-1, arrestin, beta 1, betaArrestin 1, beta-arrestin-1 | |
ARRB1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
P49407 | |
408 | |
E. coli-derived recombinant human beta-Arrestin 1 Met1-Arg418 Accession # P49407 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human beta-Arrestin 1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human beta-Arrestin 2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title