Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-Defensin 3 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | beta-Defensin 3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
beta-Defensin 3 Polyclonal specifically detects beta-Defensin 3 in Human samples. It is validated for Western Blot.Specifications
beta-Defensin 3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
BD14, BD3, BD-3, beta-defensin 103, betaDefensin 3, Beta-defensin 3, DEFB103, DEFB103A, DEFB103B, DEFB3, DEFB-3, DEFB3DEFB103A, Defensin, beta 103, defensin, beta 103B, Defensin-like protein, hBD-3, HBD3HBP3 | |
DEFB103B | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P81534 | |
55894 | |
E. coli-derived recombinant human beta-Defensin 3 Gly23-Lys67 Accession # P81534 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human beta-Defensin 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human beta-Defensin 2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title