Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta-Defensin 4/2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF2758SP
This item is not returnable.
View return policy
Description
beta-Defensin 4/2 Polyclonal specifically detects beta-Defensin 4/2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
beta-Defensin 4/2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BD-2, betaDefensin 2, Beta-defensin 2, DEFB2, DEFB-2, DEFB2DEFB102, DEFB4A, DEFB4B, DEFB4beta-defensin 4A, Defensin, beta 2Skin-antimicrobial peptide 1, defensin, beta 4, defensin, beta 4A, HBD-2, SAP-1, SAP1BD-2 | |
Goat | |
Affinity Purified | |
RUO | |
1673 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
O15263 | |
DEFB4A | |
E. coli-derived recombinant human beta-Defensin 4/2 Gly24-Pro64 Accession # O15263 | |
25 μg | |
Primary | |
Detects human beta-Defensin 4/2 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction