Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human beta IG-H3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Beta Ig-h3/TGFBI |
---|---|
Clone | 348506 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Beta Ig-h3/TGFBI Monoclonal specifically detects Beta Ig-h3/TGFBI in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
Beta Ig-h3/TGFBI | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta IGH3, Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, TGFBIP, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3 | |
TGFBI | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
348506 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q15582 | |
7045 | |
Mouse myeloma cell line NS0-derived recombinant human beta IG-H3 Gly24-His683 Accession # Q15582 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human beta IG-H3 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse beta IG-H3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title