Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human beta-NGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB256500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB256500 500 μg
MAB256SP 25 μg
MAB256100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB256500 Supplier R&D Systems Supplier No. MAB256500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

beta-NGF Monoclonal specifically detects beta-NGF in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen beta-NGF
Applications Neutralization, ELISA
Classification Monoclonal
Clone 25623
Conjugate Unconjugated
Dilution Neutralization 0.1-0.4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA36832
Gene Alias Beta nerve growth factor precursor, beta-nerve growth factor, betaNGF, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFB, NGFBMGC161428
Gene Symbols NGF
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human beta-NGF Ser122-Ala241 Accession # CAA36832
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4803
Test Specificity Detects human beta-NGF in ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) beta-NGF and recombinant rat (rr) beta-NGF is observed and less than 5% cross-reactivity with recombinant human (rh) BDNF, rhNT-3, and rhNT-4 is observed. This antibody neutralizes the bioactivity of rh beta-NGF, rm beta-NGF, and rr beta-NGF.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.