Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Beta-thromboglobulin ELISA Kit (Colorimetric)
SDP

Catalog No. NB036451 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
10 Kit
5 Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036451 10 Kit
NB036452 1 Kit
NB036453 5 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB036451 Supplier Novus Biologicals™ Supplier No. NBP28883110KIT
Add to Cart
Add to Cart
Request a Quote

Human Beta-thromboglobulin ELISA Kit (Colorimetric) measures Human Beta-thromboglobulin in serum, plasma, tissue homogenates

Specifications

Assay Range 0.78 ng/mL-50 ng/mL
Assay Sensitivity 0.195 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum, Tissue Homogenates
Specificity This assay has high sensitivity and excellent specificity for detection of human beta-TG. No significant cross-reactivity or interference between human beta-TG and analogues was observed.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5473
Gene Symbol PPBP
Quantity 10 Kit
Storage Requirements Store at 4°C.
Synonym Beta-TG, CTAP III, CXCL7, NAP-2, PBP, PPBP, TC1, TGB, THBGB1
Target Beta-thromboglobulin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.