Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Betacellulin/BTC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB261500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB261500 500 μg
MAB261SP 25 μg
MAB261100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB261500 Supplier R&D Systems Supplier No. MAB261500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Betacellulin/BTC
Applications ELISA
Classification Monoclonal
Clone 34362
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35070
Gene Alias betacellulin, BTC, probetacellulin
Gene Symbols BTC
Host Species Mouse
Immunogen E. coli-derived recombinant human Betacellulin Asp32-Tyr111 Accession # P35070
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 685
Test Specificity Detects human Betacellulin in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) EGF, rhHB-EGF, rhTGF-alpha , rhHRG-alpha , or rhAR under non-reducing conditions.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.