Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Bikunin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7744SP
This item is not returnable.
View return policy
Description
Bikunin Polyclonal specifically detects Bikunin in Human samples. It is validated for Western Blot.Specifications
Bikunin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AMBP | |
Mouse myeloma cell line NS0-derived recombinant human Bikunin Ala206-Asn352 Accession # P02760 | |
25 μg | |
Primary | |
Detects human Bikunin in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P02760 | |
Sheep | |
Affinity Purified | |
RUO | |
259 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction