Learn More
Human Bikunin Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Bikunin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL |
| Gene Accession No. | P02760 |
| Gene Symbols | AMBP |
| Host Species | Sheep |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human Bikunin Ala206-Asn352 Accession # P02760 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.