Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BLAME/SLAMF8 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA19072V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA19072V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA19072V100 Supplier R&D Systems Supplier No. FAB19072V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BLAME/SLAMF8 Monoclonal specifically detects BLAME/SLAMF8 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BLAME/SLAMF8
Applications Flow Cytometry
Classification Monoclonal
Clone 250014
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9P0V8
Gene Alias B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8
Gene Symbols SLAMF8
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BLAME/SLAMF8 Ala23-Asp233 Accession # Q9P0V8
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56833
Test Specificity Detects human BLAME/SLAMF8 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) SLAMF7 or rhSLAMF6 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.