Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BLAME/SLAMF8 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB19072N100UG
This item is not returnable.
View return policy
Description
BLAME/SLAMF8 Monoclonal specifically detects BLAME/SLAMF8 in Human samples. It is validated for Flow Cytometry.Specifications
BLAME/SLAMF8 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9P0V8 | |
SLAMF8 | |
Mouse myeloma cell line NS0-derived recombinant human BLAME/SLAMF8 Ala23-Asp233 Accession # Q9P0V8 | |
RUO | |
56833 | |
Human | |
IgG1 |
Flow Cytometry | |
250014 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8 | |
Mouse | |
100 μg | |
Primary | |
Detects human BLAME/SLAMF8 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) SLAMF7 or rhSLAMF6 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction