Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BLIMP1/PRDM1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC36081A
This item is not returnable.
View return policy
Description
BLIMP1/PRDM1 Monoclonal specifically detects BLIMP1/PRDM1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
BLIMP1/PRDM1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PRDM1, PR-domain zinc finger protein 1, ZNFPR1A1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
639 | |
Human | |
Purified |
Flow Cytometry | |
646702 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
O75626 | |
PRDM1 | |
E. coli-derived recombinant human BLIMP1/PRDM1 Lys667-Cys789 Accession # O75626 | |
100 Tests | |
Primary | |
Detects human BLIMP1/PRDM1 in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction