Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMAL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | BMAL1 |
---|---|
Clone | 1029536 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10560SP
![]() |
R&D Systems
MAB10560SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10560100
![]() |
R&D Systems
MAB10560100 |
100 μg |
Each for $432.50
|
|
|||||
Description
BMAL1 Monoclonal specifically detects BMAL1 in Human samples. It is validated for ELISA.Specifications
BMAL1 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
ARNTL, aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1c, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, JAP3, Member of PAS protein 3, member of PAS superfamily 3, MOP3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3, PASD3MGC47515, TIC | |
E. coli-derived recombinant human BMAL1, Met105-Glu303, Accession # O00327 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1029536 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
O00327 | |
406 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title