Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-10 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | BMP-10 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
BMP-10 Polyclonal specifically detects BMP-10 in Human samples. It is validated for Western Blot.Specifications
BMP-10 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
BMP10, BMP-10, bone morphogenetic protein 10, MGC126783 | |
BMP10 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O95393 | |
27302 | |
Mouse myeloma cell line NS0-derived recombinant human BMP-10 propeptide (R&D Systems, Catalog # 3956-BP) Ser20-Arg313 Accession # O95393 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human BMP-10 propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant human BMP-10 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title