Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMP-2 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. p-5379805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3551SP 25 μg
MAB3551100 100 μg
MAB3551500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB3551SP Supplier Novus Biologicals Supplier No. MAB3551SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 11 publications

BMP-2 Monoclonal specifically detects BMP-2 in Chlorocebus pygerythrus (Vervet Monkey)∣Hamster∣Human∣Mouse∣Rabbit samples. It is validated for Bioassay∣ELISA Capture (Matched Antibody Pair)∣Immunocytochemistry∣Immunohistochemistry∣Immunoprecipitation∣Neutralization∣Western Blot.

Specifications

Antigen BMP-2
Applications Bioassay, ELISA Capture (Matched Antibody Pair), Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone 100221
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P12643
Gene Alias BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Gene Symbols BMP2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human BMP-2 Gln283-Arg396 Accession # P12643
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 650
Test Specificity Detects human BMP-2 in ELISAs and Western blots. In Western blots, this antibody shows less than 1% cross-reactivity with recombinant human (rh) BMP-3, rhBMP-4, rhBMP-5, rhBMP-6, or rhBMP-7.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Vervet Monkey, Hamster, Human, Mouse, Rabbit
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.