Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMP-2/BMP-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB355100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB355100 100 μg
MAB355SP 25 μg
MAB355500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB355100 Supplier R&D Systems Supplier No. MAB355100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

BMP-2/BMP-4 Monoclonal specifically detects BMP-2/BMP-4 in Human samples. It is validated for Western Blot.

Specifications

Antigen BMP-2/BMP-4
Applications Western Blot
Classification Monoclonal
Clone 65529
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P12643
Gene Alias BDA2, BMP2, BMP-2, BMP2A, BMP-2A, BMP2ABone morphogenetic protein 2A, BMP2B, BMP2B1, BMP4, bone morphogenetic protein 2, MCOPS6, OFC11, SSFSC
Gene Symbols BMP2
Host Species Mouse
Immunogen E. coli-derived recombinant human BMP-2 Ala284-Arg396 Accession # P12643
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 650
Test Specificity Detects human BMP-2 and human BMP-4 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows no cross-reactivity with recombinant human (rh) BMP-5 or rhBMP-6.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.