Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMP-2/BMP-7 Heterodimer Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3229SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3229SP 25 μg
MAB3229 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3229SP Supplier R&D Systems Supplier No. MAB3229SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BMP-2/BMP-7 Heterodimer Monoclonal specifically detects BMP-2/BMP-7 Heterodimer in Human samples. It is validated for Western Blot.

Specifications

Antigen BMP-2/BMP-7 Heterodimer
Applications Western Blot
Classification Monoclonal
Clone 369505
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001191, NP_001710
Gene Symbols BMP2
Host Species Mouse
Immunogen E. coli-derived recombinant human BMP-2/BMP-7 Heterodimer Ala284-Arg396 (BMP-2), Ser293-His431 (BMP-7) Accession # NP_001191, NP_001710
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 650
Test Specificity Detects human BMP-2/BMP-7 Heterodimer in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) BMP-2 is observed, and no cross-reactivity with rhBMP-4/7, -4, -5, -6, -7, or -8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.