Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human BMP-2 DuoSet ELISA

Catalog No. DY355
Change view
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
DY355 15 96-well Plates
DY35505 5 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DY355 Supplier R&D Systems™ Supplier No. DY355
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human BMP-2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human BMP-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human BMP-2.

Specifications

Product Type ELISA Kit
Quantity 1 Kit
Species Human
Conjugate Biotin
Assay Range 46.90 to 3000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 15 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 650
Synonym BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.