Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF113
This item is not returnable.
View return policy
Description
BMP-3 Polyclonal specifically detects BMP-3 in Human samples. It is validated for Western Blot.Specifications
BMP-3 | |
Polyclonal | |
Biotin | |
P12645 | |
BMP3 | |
E. coli-derived recombinant human BMP-3 Ser346-Arg472 Accession # P12645 | |
50 μg | |
Primary | |
Detects human BMP-3 in Western blots. In this format, less than 1% cross-reactivity with recombinant human (rh) BMP-2, rhBMP-4, rhBMP-5, rhBMP-6, rhBMP-7 and rhBMP-8 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
BMP3, BMP-3, BMP3A, BMP-3A, bone morphogenetic protein 3, bone morphogenetic protein 3 (osteogenic), Bone morphogenetic protein 3A, bone morphogenetic protein-3, osteogenin | |
Goat | |
Affinity Purified | |
RUO | |
651 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction