Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMP-3b/GDF-10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1543
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1543 100 μg
AF1543SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1543 Supplier R&D Systems Supplier No. AF1543
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

BMP-3b/GDF-10 Polyclonal specifically detects BMP-3b/GDF-10 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen BMP-3b/GDF-10
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q5VSQ8
Gene Alias BIP, BMP3b, BMP-3b, bone morphogenetic protein 3B, Bone-inducing protein, GDF10, GDF-10, growth differentiation factor 10, Growth/differentiation factor 10
Gene Symbols GDF10
Host Species Goat
Immunogen E. coli-derived recombinant human BMP-3b/GDF-10 Gln369-Arg478 Accession # Q5VSQ8
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2662
Test Specificity Detects human BMP-3b/GDF-10 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) BMP-3 is observed and less than 2% cross-reactivity with rhBMP-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.