Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | BMP-4 |
---|---|
Clone | 1050731 |
Dilution | Immunocytochemistry 8 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7572SP
![]() |
R&D Systems
MAB7572SP |
25 μg |
Each for $200.50
|
|
|||||
MAB7572100
![]() |
R&D Systems
MAB7572100 |
100 μg |
Each for $514.50
|
|
|||||
Description
BMP-4 Monoclonal antibody specifically detects BMP-4 in Human samples. It is validated for ImmunocytochemistrySpecifications
BMP-4 | |
Immunocytochemistry 8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
E. coli-derived human BMP-4 protein, Lys303-Arg408, Accession # P12644 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1050731 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P12644 | |
652 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title