Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF757
This item is not returnable.
View return policy
Description
BMP-4 Polyclonal specifically detects BMP-4 in Human samples. It is validated for Western Blot.Specifications
BMP-4 | |
Polyclonal | |
Biotin | |
P12644 | |
BMP4 | |
Mouse myeloma cell line NS0-derived recombinant human BMP-4 Ser293-Arg408 Accession # P12644 | |
50 μg | |
Primary | |
Detects human BMP-4 in Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human (rh) BMP-2 is observed and 10% cross-reactivity with rhBMP-5 and rhBMP-6 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
Goat | |
Affinity Purified | |
RUO | |
652 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction