Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMP-4 MAb (Clone 66119), R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 12 publications

$200.16 - $2109.88

Specifications

Antigen BMP-4
Clone 66119
Dilution Western Blot 1 ug/mL, Neutralization 1-3 ug/mL
Applications Western Blot, Neutralization
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB757SP
GREENER_CHOICE
View Documents
R&D Systems
MAB757SP
25 μg
Each for $200.16
Only null left
Add to Cart
 
MAB757100
GREENER_CHOICE
View Documents
R&D Systems
MAB757100
100 μg
Each for $579.52
Only null left
Add to Cart
 
MAB757500
GREENER_CHOICE
View Documents
R&D Systems
MAB757500
500 μg
Each for $2,109.88
Only null left
Add to Cart
 
Description

Description

BMP-4 Monoclonal specifically detects BMP-4 in Human samples. It is validated for Western Blot, Neutralization.
Specifications

Specifications

BMP-4
Western Blot 1 ug/mL, Neutralization 1-3 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
BMP4
IgG2b
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
66119
Western Blot, Neutralization
Unconjugated
Mouse
Human
P12644
652
Mouse myeloma cell line NS0-derived recombinant human BMP-4 Ser293-Arg408 Accession # P12644
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human BMP-4 in direct ELISAs and Western blots. This antibody shows approximately 5% cross-reactivity with recombinant human BMP-2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.