Learn More
Gibco™ Human BMP-4 Recombinant Protein
Description
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute lyophilized recombinant human BMP-4 in 4 mM HCl containing 0.1% human serum albumin. Further dilutions should be made in low endotoxin medium or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA. Storage: Store this lyophilized preparation at =-20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at =-20°C. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Accession Number | P12644, P21275 |
| For Use With (Application) | Control, ELISA, Functional Assay, Western Blot |
| Formulation | HCl with no preservative |
| Gene ID (Entrez) | 12159, 652 |
| Molecular Weight (g/mol) | 34 kDa |
| Name | Human BMP-4 |
| Purification Method | Purified |
| Quantity | 100 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.