Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7151SP
This item is not returnable.
View return policy
Description
BMP-5 Monoclonal specifically detects BMP-5 in Human samples. It is validated for Neutralization.Specifications
BMP-5 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP5, BMP-5, bone morphogenetic protein 5, MGC34244 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
653 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Neutralization | |
149107 | |
Neutralization 2-10 ug/mL | |
P22003 | |
BMP5 | |
Mouse myeloma cell line NS0-derived recombinant human BMP-5 Asn313-His454 Accession # P22003 | |
25 μg | |
Primary | |
Detects human BMP-5 in direct ELISAs and Western blots. In direct ELISAs, 50% cross-reactivity with recombinant human (rh) BMP-6 is observed and no cross-reactivity with rhBMP-2, -3, -4, or -7 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction