Learn More
Description
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA Development Kit |
| Quantity | 15 x 96-well Plates |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 156.00 to 10000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 654 |
| Product Line | R&D Systems |
| Synonym | BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
