Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3542100
This item is not returnable.
View return policy
Description
BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
BMP-7 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1OP1, Osteogenic protein 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
655 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Immunohistochemistry, Neutralization, ELISA | |
164313 | |
Immunohistochemistry 8-25 ug/mL, Neutralization 4-12 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P18075 | |
BMP7 | |
Chinese hamster ovary cell line CHO-derived recombinant human BMP-7 Arg292-His431 Accession # P18075 | |
100 μg | |
Primary | |
Detects human BMP-7 in ELISAs. In ELISAs, this antibody shows approximately 25-50% cross-reactivity with recombinant human (rh) BMP-6 and no cross-reactivity with rhBMP-2, -3, -4, -5, or -8. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction