Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMP-7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3541100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3541100 100 μg
MAB3541SP 25 μg
MAB3541500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB3541100 Supplier R&D Systems Supplier No. MAB3541100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen BMP-7
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 164311
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 1.5-6.0 ug/mL
Formulation Supplied as a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P18075
Gene Alias BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1OP1, Osteogenic protein 1
Gene Symbols BMP7
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human BMP-7 Ser293-His431 Accession # P18075
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 655
Test Specificity Detects human BMP-7 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) BMP-6 is observed and no cross-reactivity with rhBMP-2, -3, -4, -5, or -8 is observed.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.