Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | BMP-8 |
---|---|
Clone | 158708 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
BMP-8 Monoclonal specifically detects BMP-8 in Human samples. It is validated for Western Blot.Specifications
BMP-8 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
AAP74559 | |
353500 | |
E. coli-derived recombinant human BMP-8 Ala264-His402 Accession # AAP74559 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human BMP-8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human BMP-2, -3, -4, -5, -6, or -7 is observed. |
158708 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP8, BMP8A, BMP-8A, OP-2 | |
BMP8A | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title