Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-8 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1073
This item is not returnable.
View return policy
Description
BMP-8 Polyclonal specifically detects BMP-8 in Human samples. It is validated for Western Blot.Specifications
BMP-8 | |
Polyclonal | |
Biotin | |
AAP74559 | |
BMP8A | |
E. coli-derived recombinant human BMP-8a (R&D Systems, Catalog # 1073-BP) Ala264-His402 Accession # AAP74559 | |
50 μg | |
Primary | |
Detects human BMP-8 in Western blots. In this format, less than 1% cross-reactivity with recombinant human (rh) BMP-2, rhBMP-3, rhBMP-4, rhBMP-5, rhBMP-6 and rhBMP-7 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
BMP8, BMP8A, BMP-8A, OP-2 | |
Goat | |
Affinity Purified | |
RUO | |
353500 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction